Please login to your online account to display your discounted pricing

Pirfenidone, Tocris

Antifibrotic agent; regulates cytokine levels in vivo

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material Pirfenidone
Assay Percent Range >99%
Molecular Formula C12H11NO
Formula Weight 185.23
Synonym 5-methyl-1-phenyl-2(1H)-pyridinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

109310

 
r & d systems
RDS109310
10mg Each for $168.15

109350

 
r & d systems
RDS109350
50mg Each for $697.40
Description & Specifications

Specifications

Formula Weight 185.23
Quantity 10mg
Chemical Name or Material Pirfenidone
Assay Percent Range >99%
Molecular Formula C12H11NO
CAS 53179-13-8
Synonym 5-methyl-1-phenyl-2(1H)-pyridinone
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature