Please login to your online account to display your discounted pricing

Pirfenidone, Tocris


Antifibrotic agent; regulates cytokine levels in vivo

Quantity 10mg

Manufacturer: r & d systems  RDS109310

 View more versions of this product

Catalog No. 109310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 185.23
Quantity 10mg
Assay Percent Range >99%
CAS 53179-13-8
Synonym 5-methyl-1-phenyl-2(1H)-pyridinone
Chemical Name or Material Pirfenidone
Recommended Storage Store at Room Temperature
Molecular Formula C12H11NO

Specifications

Chemical Name or Material Pirfenidone
Assay Percent Range >99%
Molecular Formula C12H11NO
Formula Weight 185.23
Synonym 5-methyl-1-phenyl-2(1H)-pyridinone
Accessories

Store at Room Temperature