Please login to your online account to display your discounted pricing

Tocris™ PIR 3.5

Negative control of IPA 3 (Cat. No. 3622)

$598.33 - $598.33

Specifications

Product Type PIR 3.5
Inhibitors PIR 3.5
Molecular Weight 350.45
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

421250

 
tocris™
RDS421250
50mg Each for $598.33
Description & Specifications

Specifications

Product Type PIR 3.5
Inhibitors PIR 3.5
Molecular Weight 350.45
Purity >98

Negative control of IPA 3 (Cat. No. 3622) a direct, non-competitive inhibitor of group I p21-activated kinase (Pak1).

  • Chemical Name: 6,6'-Dithiodi-2-naphthtol
  • Formula: C20H14O2S2
  • CAS Number: 6088-51-3
  • Solubility: Soluble to 75mM in DMSO and to 10mM in ethanol
  • Purity: >98