Please login to your online account to display your discounted pricing

Pipsyl chloride, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  207290050

 View more versions of this product

Catalog No. AC207290050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (ex Cl)
Formula Weight 302.51
Physical Form Crystals or Crystalline Powder
Quantity 5g
Color White to beige or orange
Beilstein 11, 65
Merck Index 15, 7599
EINECS Number 202-686-9
Melting Point 80° to 84°C
TSCA TSCA
Synonym 4-Iodobenzenesulfonyl chloride
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 97%
Chemical Name or Material Pipsyl chloride
MDL Number MFCD00007441
Name Note 97%
Molecular Formula C6H4ClIO2S
CAS 98-61-3
Linear Formula IC6H4SO2Cl

Specifications

Chemical Name or Material Pipsyl chloride
Name Note 97%
CAS 98-61-3
Assay ≥96.0% (ex Cl)
Assay Percent Range 97%
Molecular Formula C6H4ClIO2S
Linear Formula IC6H4SO2Cl
Formula Weight 302.51
MDL Number MFCD00007441
Synonym 4-Iodobenzenesulfonyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents