Please login to your online account to display your discounted pricing

PIPPS, Calbiochem™, EMD Millipore

A non-metal complexing, zwitterionic buffer with mixed-mode dissociation constants of pKa1 = 3.73 and pKa2 = 7.96 (100mM, 25°C)

$299.00 - $299.00

Chemical Identifiers

Assay Percent Range ≥97.0%
Formula Weight 330.4
CAS 5625-56-9
Chemical Name or Material PIPPS
Molecular Formula C10H22N2O6S2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

528315100GM

 
emd millipore corporation
528315100GM
Each for $299.00
Description & Specifications

Specifications

Formula Weight 330.4
Quantity 100g
Chemical Name or Material PIPPS
Assay Percent Range ≥97.0%
Molecular Formula C10H22N2O6S2
CAS 5625-56-9
Synonym Piperazine-N,N'-bis(3-propanesulfonic Acid)
Solubility Information Soluble in 0.25N NaOH solution or water
Recommended Storage 2°C to 8°C (Do not freeze).

PIPPS has no interference at wavelengths longer than 240nm

Safety and Handling

2°C to 8°C (Do not freeze).