Please login to your online account to display your discounted pricing

Gilson™ PIPETMAN M Single Channel Pipets


Fully motorized, lightweight pipet that virtually eliminates pipetting forces and minimizes RSI (Repetitive Strain Injuries) among users.

Manufacturer: gilson™  F81022

 View more versions of this product

Catalog No. F81022G

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) D10, DL10, DF10ST, DFL10ST pipette tip
Includes Fully motorized piston, color coded push button, universal tip holder
Model P10M
Volume (Metric) 0.5 to 10μL
Number of Channels 1

An ergonomic pipetting solution with GLP features that each lab needs

  • Motorized piston with adjustable speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes – reverse, mix, repetitive and pipetting – to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.