Please login to your online account to display your discounted pricing
Gilson™ PIPETMAN™ Diamond™ Pipette Tips - Reload Packs
RELOAD PACK is easiest and quickest way to refill empty TIPACK Box
$50.63 - $50.63
Specifications
Product Type Reload Pack
View More Specs
Products
Catalog Number Mfr. No. Model Volume (Metric) Range Price Quantity    
F167022G
gilson™
F167012
DL10 0.1 - 20μL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
F167024G
gilson™
F167014
D1000 100 - 1000μL Pack of 576 Each for $50.63
Description & Specifications
Specifications
Product Type Reload Pack
  • Three times smaller than regular tips box
  • Ecofriendly - uses up to 70% less plastic than regular rack box
  • Color-coded racks to differentiate tip models and indicate compatible Gilson pipette models
  • Autoclaving conditions: Moist heat/121°C/20 minutes/1 bar