Gilson™ PIPETMAN™ Diamond™ Pipette Tips - Reload Packs
RELOAD PACK is easiest and quickest way to refill empty TIPACK Box
Manufacturer:  Gilson™ F167012
 View more versions of this product
Catalog No. F167022G
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Pack, Reload; Gilson; PIPETMAN; Diamond; Model: DL10; For refilling empty TIPACK box; Easy to store on all benches; Color-coded racks; Autoclavable; Vol. Range: 0.1 - 20μL; 10 racks x 96 tips
  • Three times smaller than regular tips box
  • Ecofriendly - uses up to 70% less plastic than regular rack box
  • Color-coded racks to differentiate tip models and indicate compatible Gilson pipette models
  • Autoclaving conditions: Moist heat/121°C/20 minutes/1 bar
Description & Specifications
Specifications
DL10
Reload Pack
0.1 - 20μL