Please login to your online account to display your discounted pricing

PIPES, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$157.84 - $448.90

Chemical Identifiers

Chemical Name or Material PIPES
CAS 5625-37-6
Purity Grade Ultrapure Bioreagent
Molecular Formula C8H18N2O6S2
Formula Weight 302.37
Synonym piperazine-N,N'-bis(2-ethanesulfonic acid)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003623

 
avantor performance
426501
Poly Bottle 100g Each for $157.84

02003625

 
avantor performance
426500
POLYSTORMOR 25g Each for $448.90
Description & Specifications

Specifications

Formula Weight 302.37
Molecular Formula C8H18N2O6S2
CAS 5625-37-6
Chemical Name or Material PIPES
Purity Grade Ultrapure Bioreagent
Synonym piperazine-N,N'-bis(2-ethanesulfonic acid)
Packaging Poly Bottle
Quantity 100g