Please login to your online account to display your discounted pricing

PIPES sesquisodium salt, 97%, for biochemistry, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  327781000

 View more versions of this product

Catalog No. AC327781000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96%
Physical Form Powder
Formula Weight 335.34
Quantity 100g
Color White
pH 6.5 to 7.1 (1% soln. at 25°C)
Purity Grade For biochemistry
Absorbance (10% aq. soln.) (1 cm cell)
Water 4% max. (K.F.)
Chemical Name or Material PIPES sesquisodium salt
Absorbance Conditions (10% aq. soln.) (1 cm cell)
Name Note For biochemistry
Assay Percent Range 97%
Molecular Formula C16H33N4Na3O12S4
MDL Number MFCD00066393
CAS 100037-69-2
Synonym Piperazine-N, N'-bis(2-ethanesulfonic acid) sesquisodium salt
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material PIPES sesquisodium salt
Name Note For biochemistry
CAS 100037-69-2
Purity Grade For biochemistry
Assay ≥96%
Assay Percent Range 97%
Molecular Formula C16H33N4Na3O12S4
Formula Weight 335.34
MDL Number MFCD00066393
Synonym Piperazine-N, N'-bis(2-ethanesulfonic acid) sesquisodium salt