Please login to your online account to display your discounted pricing

Alfa Aesar™ PIPES monosodium salt hydrate, 99%

$174.78 - $174.78

Chemical Identifiers

Chemical Name or Material PIPES monosodium salt hydrate
CAS 10010-67-0
Molecular Formula C8H17N2NaO6S2·xH2O
Formula Weight 324.34 (Anhydrous)
MDL Number MFCD00065472
Merck Index 14,7479
Synonym Piperazine-1,4-bis(ethanesulfonic acid) monosodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2183522

  SDS
alfa aesar™
B2183522
100g Each for $174.78
Description & Specifications

Specifications

Formula Weight 324.34 (Anhydrous)
Synonym Piperazine-1,4-bis(ethanesulfonic acid) monosodium salt
CAS 10010-67-0
MDL Number MFCD00065472
Chemical Name or Material PIPES monosodium salt hydrate
Assay 99%
Merck Index 14,7479
TSCA TSCA
Melting Point >300°C
Molecular Formula C8H17N2NaO6S2·xH2O
Quantity 100g

Applications
PIPES [piperazine-N,N′-bis(2-ethanesulfonic acid)] is frequently used as a buffering agent in biochemistry. It is useful in cell culture work. It has been documented to minimize lipid loss when buffering glutaraldehyde histology in plant and animal tissues.

Solubility
Soluble in water

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.