Please login to your online account to display your discounted pricing

PIPES, Free Acid, ULTROL™ Grade, Calbiochem™, EMD Millipore

A zwitterionic N-substituted aminosulfonic acid useful in the pH range of 6.1 to 7.5

$107.35 - $107.35

Chemical Identifiers

Assay Percent Range ≥98.0%
Formula Weight 302.4
CAS 5625-37-6
Chemical Name or Material PIPES, Free Acid
Molecular Formula C8H18N2O6S2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

528131100GM

 
emd millipore corporation
528131100GM
Each for $107.35
Description & Specifications

Specifications

Formula Weight 302.4
Quantity 100g
Recommended Storage 15°C to 30°C (Do not freeze).
CAS 5625-37-6
Purity ULTROL™ Grade
Molecular Formula C8H18N2O6S2
Assay Percent Range ≥98.0%
Chemical Name or Material PIPES, Free Acid
Solubility Information Soluble in water or aqueous buffer
Purity Grade Notes by titration (dry basis)
Synonym Piperazine-N,N'-bis(2-ethanesulfonic Acid)
Safety and Handling

15°C to 30°C (Do not freeze).