Please login to your online account to display your discounted pricing

PIPES, free acid, MP Biomedicals™


Quantity 100g

Manufacturer: mp biomedicals  19455280

 View more versions of this product

Catalog No. ICN19455280

  • / Each
 

Description & Specifications

Chemical Identifiers

Color White
Purity Grade Cell Culture Reagent Grade
Physical Form Crystalline Powder
Formula Weight 302.4
Quantity 100g
EINECS Number 227-057-6
Chemical Name or Material PIPES, free acid
MDL Number MFCD00006159
CAS 5625-37-6
Synonym 1, 4-Piperazinediethanesulfonic acid
Molecular Formula C8H18N2O6S2

Specifications

Chemical Name or Material PIPES, free acid
CAS 5625-37-6
Purity Grade Cell Culture Reagent Grade
Physical Form Crystalline Powder
Molecular Formula C8H18N2O6S2
Formula Weight 302.4
MDL Number MFCD00006159
Synonym 1, 4-Piperazinediethanesulfonic acid