missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PIPES, disodium salt, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 215090250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
0.050 max. (0.1M in water) at 260nm, 0.050 max. (0.1M in water) at 280nm, 0.050 max. (260nm, 0.1M in water) | |
76836-02-7 | |
9.2 to 10.0 max. (25°C, 1% min. aq. soln.) | |
25 g | |
Authentic | |
Glass bottle | |
GMHSTJRPSVFLMT-UHFFFAOYSA-L | |
disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate | |
173553 | |
346.33 |
PIPES, disodium salt | |
White | |
Crystalline Powder or Powder | |
99% | |
C8H16N2Na2O6S2 | |
1, 4-Piperazinediethanesulfonic acid disodium salt | |
C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+] | |
346.324 | |
CHEBI:63055 | |
99% |
Chemical Identifiers
76836-02-7 | |
346.324 | |
1, 4-Piperazinediethanesulfonic acid disodium salt | |
CHEBI:63055 | |
C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+] |
C8H16N2Na2O6S2 | |
GMHSTJRPSVFLMT-UHFFFAOYSA-L | |
173553 | |
disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate |
RUO – Research Use Only