missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ PIPES, 0.5M buffer soln., pH 6.0

$59.51 - $93.45

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62224AE View Documents Alfa Aesar™
J62224AE
100mL Each for $59.51
 
AAJ62224AK View Documents Alfa Aesar™
J62224AK
250mL Each for $93.45
 
Specifications

Chemical Identifiers

5625-37-6
302.36
IHPYMWDTONKSCO-UHFFFAOYSA-N
CHEBI:44933
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
C8H18N2O6S2
MFCD00006159
79723
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid

Specifications

5625-37-6
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
79723
Liquid
Colorless
PIPES buffer soln.
C8H18N2O6S2
0.5M buffer soln.
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
302.36
CHEBI:44933
6.0
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C