Please login to your online account to display your discounted pricing

Alfa Aesar™ PIPES, 0.2M buffer soln., pH 6.8, low endotoxin

Chemical Identifiers

Chemical Name or Material PIPES
CAS 5625-37-6
Name Note low endotoxin
MDL Number MFCD00006159
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67229AE

 
alfa aesar™
J67229AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67229AK

 
alfa aesar™
J67229AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material PIPES
CAS 5625-37-6
Physical Form Liquid
pH 6.8
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Concentration 0.2M buffer soln.
MDL Number MFCD00006159
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C