Please login to your online account to display your discounted pricing

Alfa Aesar™ PIPES, 0.2M buffer soln., pH 6.5, low endotoxin


Quantity 100mL

Manufacturer: alfa aesar™  J67491AE

 View more versions of this product

Catalog No. AAJ67491AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material PIPES
MDL Number MFCD00006159
pH 6.5
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note low endotoxin
Physical Form Liquid
Concentration 0.2M buffer soln.
CAS 5625-37-6
Storage Note 1 2° to 8°C

Specifications

Chemical Name or Material PIPES
CAS 5625-37-6
Name Note low endotoxin
MDL Number MFCD00006159
Accessories

2° to 8°C