missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ PIPES, 0.2M buffer soln., pH 6.5, low endotoxin

$64.00 - $106.26

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67491AE View Documents Alfa Aesar™
J67491AE
100mL Each for $64.00
 
AAJ67491AK View Documents Alfa Aesar™
J67491AK
250mL Each for $106.26
 
Specifications

Chemical Identifiers

5625-37-6
302.36
IHPYMWDTONKSCO-UHFFFAOYSA-N
CHEBI:44933
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
C8H18N2O6S2
MFCD00006159
79723
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid

Specifications

5625-37-6
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
79723
Liquid
Colorless
PIPES, low endotoxin
C8H18N2O6S2
0.2M buffer soln.
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
302.36
CHEBI:44933
6.5
100mL
Safety and Handling

Safety and Handling

TSCA : TSCA

storageNote1 : 2° to 8°C