missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Piperonyloyl chloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC370790100

 View more versions of this product

Catalog No. AC370790100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
184.58
Off-white to blue-gray
Authentic
98%
Glass bottle
C8H5ClO3
Crystalline Powder
10g
77° to 81°C
MFCD00016904
25054-53-9
3, 4-(Methylenedioxy)benzoyl chloride
Piperonyloyl chloride

Chemical Identifiers

25054-53-9
C8H5ClO3
MFCD00016904
Piperonyloyl chloride
98%
184.58
3, 4-(Methylenedioxy)benzoyl chloride
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger