Please login to your online account to display your discounted pricing

Piperonylonitrile 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  151881000

 View more versions of this product

Catalog No. AC151881000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Needles
Assay ≥96.0%
Formula Weight 147.13
Quantity 100g
Color White to beige
Beilstein 19, 270
EINECS Number 224-590-6
Melting Point 90° to 95°C
RTECS Number TO2645000
Chemical Name or Material Piperonylonitrile
Name Note 97%
Assay Percent Range 97%
Infrared Spectrum Authentic
Molecular Formula C8H5NO2
MDL Number MFCD00005820
CAS 4421-09-4
Synonym 3, 4-(Methylenedioxy)benzonitrile
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Piperonylonitrile
Name Note 97%
CAS 4421-09-4
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H5NO2
Formula Weight 147.13
MDL Number MFCD00005820
Synonym 3, 4-(Methylenedioxy)benzonitrile
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents