Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperonylonitrile, 97%

Chemical Identifiers

Chemical Name or Material Piperonylonitrile
CAS 4421-09-4
Molecular Formula C8H5NO2
Formula Weight 147.13
MDL Number MFCD00005820
UN Number UN3439
Beilstein 6224
Synonym 1,3-Benzodioxole-5-carbonitrile; 3,4-(Methylenedioxy)benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1301206

 
alfa aesar™
A1301206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1301214

 
alfa aesar™
A1301214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1301222

 
alfa aesar™
A1301222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.13
Chemical Name or Material Piperonylonitrile
Assay 97%
Molecular Formula C8H5NO2
MDL Number MFCD00005820
CAS 4421-09-4
UN Number UN3439
Beilstein 6224
Synonym 1,3-Benzodioxole-5-carbonitrile; 3,4-(Methylenedioxy)benzonitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 92° to 94°C
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECS Number TO2645000
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.