Learn More
Piperonylic acid hydrazide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5071306
Description
Piperonylic acid hydrazide is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPiperonylic acid hydrazide is used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Piperonylic acid hydrazide | |
176°C to 178°C | |
MFCD00060505 | |
3,4-methylenedioxybenzhydrazide, benzo d 1,3 dioxole-5-carbohydrazide, piperonylic acid hydrazide, 2h-1,3-benzodioxole-5-carbohydrazide, 3,4-methylenedioxybenzoic acid hydrazide, 3,4-methylenedioxy benzohydrazide, 1,3-benzodioxole-5-carboxylic acid, hydrazide, 3,4-methylenedioxy benzhydrazide, benzo 1,3 dioxole-5-carboxylic acid hydrazide, 2h-benzo d 1,3-dioxolane-5-carbohydrazide | |
RAXBGBHBUFGWPG-UHFFFAOYSA-N | |
1,3-benzodioxole-5-carbohydrazide | |
89158 | |
97% |
22026-39-7 | |
C8H8N2O3 | |
5 g | |
Slightly soluble in water. | |
C1OC2=C(O1)C=C(C=C2)C(=O)NN | |
180.163 | |
180.16 |
Chemical Identifiers
22026-39-7 | |
180.163 | |
RAXBGBHBUFGWPG-UHFFFAOYSA-N | |
89158 | |
C1OC2=C(O1)C=C(C=C2)C(=O)NN |
C8H8N2O3 | |
MFCD00060505 | |
3,4-methylenedioxybenzhydrazide, benzo d 1,3 dioxole-5-carbohydrazide, piperonylic acid hydrazide, 2h-1,3-benzodioxole-5-carbohydrazide, 3,4-methylenedioxybenzoic acid hydrazide, 3,4-methylenedioxy benzohydrazide, 1,3-benzodioxole-5-carboxylic acid, hydrazide, 3,4-methylenedioxy benzhydrazide, benzo 1,3 dioxole-5-carboxylic acid hydrazide, 2h-benzo d 1,3-dioxolane-5-carbohydrazide | |
1,3-benzodioxole-5-carbohydrazide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only