Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperonyl isobutyrate, 99%

$44.76 - $146.30

Chemical Identifiers

Chemical Name or Material Piperonyl isobutyrate
CAS 5461-08-5
Molecular Formula C12H14O4
Formula Weight 222.24
MDL Number MFCD00005826
Synonym Isobutyric acid piperonyl ester; 3,4-Methylenedioxybenzyl isobutyrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2132114

  SDS
alfa aesar™
B2132114
25g Each for $44.76

AAB2132122

  SDS
alfa aesar™
B2132122
100g Each for $146.30
Description & Specifications

Specifications

Formula Weight 222.24
Chemical Name or Material Piperonyl isobutyrate
TSCA TSCA
Assay 99%
Molecular Formula C12H14O4
MDL Number MFCD00005826
CAS 5461-08-5
Synonym Isobutyric acid piperonyl ester; 3,4-Methylenedioxybenzyl isobutyrate
Boiling Point 91° to 92°C (0.005mmHg)
Quantity 25g
Flash Point >110°C (230°F)