missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Piperonyl butoxide, 90%, Tech., Thermo Scientific™
Supplier: Thermo Scientific Chemicals 334165000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Piperonyl butoxide | |
51-03-6 | |
180°C (1.0mmHg) | |
Authentic | |
Glass bottle | |
1.4970 to 1.507 | |
500mL | |
1.06 | |
piperonyl butoxide, butacide, butocide, ethanol butoxide, pyrenone 606, piperonylbutoxide, 6-propylpiperonyl butyl diethylene glycol ether, butyl carbitol 6-propylpiperonyl ether, 5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole, butoxide synergist | |
FIPWRIJSWJWJAI-UHFFFAOYSA-N | |
5-[2-(2-butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole | |
5794 | |
CHEBI:32687 | |
90% |
tech., 90% | |
1.0600g/mL | |
170°C | |
88% min. (GC) | |
C19H30O5 | |
MFCD00005842 | |
19, IV, 779 | |
15, 7589 | |
Solubility in water: practically insoluble. Other solubilities: miscible with methanol,ethanol,benzene,freons,,and other organic solvents | |
CCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC | |
338.44 | |
40 mPa.s (25°C) | |
338.44 | |
Technical |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 200-076-7
RTECSNumber : XS8050000
TSCA : TSCA
RUO â Research Use Only