Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperonaldoxime, 99%

$79.24 - $304.36

Chemical Identifiers

Chemical Name or Material Piperonaldoxime
CAS 2089-36-3
Molecular Formula C8H7NO3
Formula Weight 165.15
MDL Number MFCD00016905
Beilstein 83661
Synonym 1,3-Benzodioxole-5-carboxaldoxime; 3,4-(Methylenedioxy)benzaldoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1195206

  SDS
alfa aesar™
L1195206
5g Each for $79.24

AAL1195214

  SDS
alfa aesar™
L1195214
25g Each for $304.36
Description & Specifications

Specifications

Formula Weight 165.15
Assay 99%
Molecular Formula C8H7NO3
MDL Number MFCD00016905
CAS 2089-36-3
Beilstein 83661
Quantity 5g
RTECS Number TO1950000
Chemical Name or Material Piperonaldoxime
Synonym 1,3-Benzodioxole-5-carboxaldoxime; 3,4-(Methylenedioxy)benzaldoxime
Melting Point 114° to 116°C