missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperonaldoxime, 99%

Manufacturer:  Alfa Aesar™ L1195214

 View more versions of this product

Catalog No. AAL1195214



Specifications

Specifications

Piperonaldoxime
25g
C8H7NO3
83661
VDAJDWUTRXNYMU-RUDMXATFSA-N
(NE)-N-(1,3-benzodioxol-5-ylmethylidene)hydroxylamine
6861467
99%
114°C to 116°C
2089-36-3
MFCD00016905
piperonaldoxime, piperonal, oxime, 3,4-methylenedioxybenzaldoxime, piperonal, syn-oxime, 1,3-benzodioxole-5-carboxaldehyde, oxime, ne-n-1,3-benzodioxol-5-ylmethylidene hydroxylamine, 3,4-methylenedioxybenzaldehyde oxime, e-benzo d 1,3 dioxole-5-carbaldehyde oxime, 1-1,3-benzodioxol-5-yl-n-hydroxymethanimine, n-2h-1,3-benzodioxol-5-ylmethylidene hydroxylamine
C1OC2=C(O1)C=C(C=C2)C=NO
165.148
165.15

Chemical Identifiers

2089-36-3
165.148
VDAJDWUTRXNYMU-RUDMXATFSA-N
6861467
C1OC2=C(O1)C=C(C=C2)C=NO
C8H7NO3
MFCD00016905
piperonaldoxime, piperonal, oxime, 3,4-methylenedioxybenzaldoxime, piperonal, syn-oxime, 1,3-benzodioxole-5-carboxaldehyde, oxime, ne-n-1,3-benzodioxol-5-ylmethylidene hydroxylamine, 3,4-methylenedioxybenzaldehyde oxime, e-benzo d 1,3 dioxole-5-carbaldehyde oxime, 1-1,3-benzodioxol-5-yl-n-hydroxymethanimine, n-2h-1,3-benzodioxol-5-ylmethylidene hydroxylamine
(NE)-N-(1,3-benzodioxol-5-ylmethylidene)hydroxylamine
Safety and Handling

Safety and Handling

RTECSNumber : TO1950000

Documents