Please login to your online account to display your discounted pricing

Piperlongumine, Tocris™

Induces apoptosis; increases p53 and ROS levels in cancer cells

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material Piperlongumine
Assay Percent Range >97%
Molecular Formula C17H19NO5
Formula Weight 317.34
Synonym 5,6-Dihydro-1-[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propen-1-yl]-2(1H)-pyridinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

439610

 
r & d systems
RDS439610
10mg Each for $214.12

439650

 
r & d systems
RDS439650
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 317.34
Quantity 10mg
Chemical Name or Material Piperlongumine
Molecular Formula C17H19NO5
CAS 20069-09-4
Recommended Storage Store at -20°C
Assay Percent Range >97%
Synonym 5,6-Dihydro-1-[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propen-1-yl]-2(1H)-pyridinone
Safety and Handling

Store at -20°C