missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Piperlongumine, TCI America™

Manufacturer:  TCI America P2361100MG

 View more versions of this product

Catalog No. P2361100MG



Specifications

Specifications

Piperlongumine
125°C
20069-09-4
C17H19NO5
5,6-Dihydro-1-[1-oxo-3-(3,4,5-trimethoxyphenyl)-trans-2-propenyl]-2(1H)-pyridinone, (E)-1-[3-(3,4,5-Trimethoxyphenyl)acryloyl]-5,6-dihydropyridin-2(1H)-one
COC1=CC(=CC(=C1OC)OC)C=CC(=O)N2CCC=CC2=O
317.341
CHEBI:8241
Crystal-Powder at 20°C
White-Yellow
100mg
≥97.0% (HPLC)
MFCD00075706
VABYUUZNAVQNPG-BQYQJAHWSA-N
1-[(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]-2,3-dihydropyridin-6-one
637858
317.34

Chemical Identifiers

20069-09-4
317.341
VABYUUZNAVQNPG-BQYQJAHWSA-N
637858
1-[(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]-2,3-dihydropyridin-6-one
C17H19NO5
MFCD00075706
5,6-Dihydro-1-[1-oxo-3-(3,4,5-trimethoxyphenyl)-trans-2-propenyl]-2(1H)-pyridinone, (E)-1-[3-(3,4,5-Trimethoxyphenyl)acryloyl]-5,6-dihydropyridin-2(1H)-one
CHEBI:8241
COC1=CC(=CC(=C1OC)OC)C=CC(=O)N2CCC=CC2=O
Safety and Handling

Safety and Handling

RTECSNumber : UU7785850

TSCA : No

Recommended Storage : Refrigerator

Documents