Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperidine-1-thiocarboxamide, 98%

$88.05 - $88.05

Chemical Identifiers

Chemical Name or Material Piperidine-1-thiocarboxamide
CAS 14294-09-8
Molecular Formula C6H12N2S
Formula Weight 144.24
MDL Number MFCD00041230
Beilstein 383640
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1244503

 
alfa aesar™
L1244503
1g Each for $88.05
Description & Specifications

Specifications

Formula Weight 144.24
Chemical Name or Material Piperidine-1-thiocarboxamide
Assay 98%
Molecular Formula C6H12N2S
MDL Number MFCD00041230
CAS 14294-09-8
Beilstein 383640
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 126° to 130°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.