missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperidine-1-thiocarboxamide, 98%

Manufacturer:  Alfa Aesar™ L1244506

 View more versions of this product

Catalog No. AAL1244506



Specifications

Specifications

Piperidine-1-thiocarboxamide
5g
C6H12N2S
383640
UERQMZVFUXRQOD-UHFFFAOYSA-N
piperidine-1-carbothioamide
1551232
98%
126°C to 130°C
14294-09-8
MFCD00041230
1-piperidinethiocarboxamide, 1-piperidinecarbothioamide, aminopiperidylmethane-1-thione, acmc-20aos3, piperidine thiocarboxamide, 1-thiocarbamyl piperidine, 1-piperidinethiocarboximide, 1-piperi-dinecarbothioamide, piperidine-1-thiocarboxamide, 1-piperidinecarbothioamide #
C1CCN(CC1)C(=S)N
144.236
144.24

Chemical Identifiers

14294-09-8
144.236
UERQMZVFUXRQOD-UHFFFAOYSA-N
1551232
C1CCN(CC1)C(=S)N
C6H12N2S
MFCD00041230
1-piperidinethiocarboxamide, 1-piperidinecarbothioamide, aminopiperidylmethane-1-thione, acmc-20aos3, piperidine thiocarboxamide, 1-thiocarbamyl piperidine, 1-piperidinethiocarboximide, 1-piperi-dinecarbothioamide, piperidine-1-thiocarboxamide, 1-piperidinecarbothioamide #
piperidine-1-carbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents