Please login to your online account to display your discounted pricing

piperidine-1-carboximidamide hydroiodide, Maybridge

Chemical Identifiers

Chemical Name or Material Piperidine-1-carboximidamide hydroiodide
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H13N3·HI
Formula Weight 255.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM00642DA

 
fisher scientific company
KM00642DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 255.1
Quantity 1g
Assay 97%
Chemical Name or Material Piperidine-1-carboximidamide hydroiodide
Assay Percent Range 97%
Molecular Formula C6H13N3·HI
CAS 102392-91-6