Please login to your online account to display your discounted pricing

piperidine-1-carboximidamide hydroiodide, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  KM00642DA

 View more versions of this product

Catalog No. KM00642DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 255.1
Quantity 1g
Assay 97%
Chemical Name or Material Piperidine-1-carboximidamide hydroiodide
Molecular Formula C6H13N3·HI
Assay Percent Range 97%
CAS 102392-91-6

Specifications

Chemical Name or Material Piperidine-1-carboximidamide hydroiodide
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H13N3·HI
Formula Weight 255.1