missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperazine hydrogen phosphate monohydrate, 98+%

Manufacturer:  Alfa Aesar™ A114870E

 View more versions of this product

Catalog No. AAA114870E


 

Specifications

Specifications

Piperazine hydrogen phosphate monohydrate
18534-18-4
C4H15N2O5P
PMGABIJVFLPSLS-UHFFFAOYSA-N
phosphoric acid;piperazine;hydrate
443965
202.14 (184.12 Anhydrous)
2500g
≥98%
MFCD00156159
C1CNCCN1.O.OP(=O)(O)O
202.147
CHEBI:32006

Chemical Identifiers

18534-18-4
202.147
PMGABIJVFLPSLS-UHFFFAOYSA-N
CHEBI:32006
C4H15N2O5P
MFCD00156159
443965
SDS Documents