Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperazine hydrogen phosphate monohydrate, 98+%

$200.41 - $200.41

Chemical Identifiers

Chemical Name or Material Piperazine hydrogen phosphate monohydrate
CAS 18534-18-4
Molecular Formula C4H10N2·H3PO4·H2O
Formula Weight 202.14 (184.12 Anhydrous)
MDL Number MFCD00156159
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1148736

  SDS
alfa aesar™
A1148736
500g Each for $200.41
Description & Specifications

Specifications

Formula Weight 202.14 (184.12 Anhydrous)
Chemical Name or Material Piperazine hydrogen phosphate monohydrate
Assay ≥98%
MDL Number MFCD00156159
Quantity 500g
Molecular Formula C4H10N2·H3PO4·H2O
CAS 18534-18-4