missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperazine hydrate, ≥97%

Manufacturer:  Alfa Aesar™ H3706930

 View more versions of this product

Catalog No. AAH3706930



Specifications

Specifications

Piperazine hydrate
125°C to 130°C
42°C to 46°C
11 to 12
C4H22N2O6
UN2579
AVRVZRUEXIEGMP-UHFFFAOYSA-N
piperazine;hexahydrate
120181
Crystalline Powder or Flakes
Weak, characteristic
White
250g
142-63-2
MFCD00149389
piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, piperazine hexahydrate, vermyl tn
C1CNCCN1.O.O.O.O.O.O
194.228
86.14 (Anhydrous)
≥97%

Chemical Identifiers

142-63-2
194.228
AVRVZRUEXIEGMP-UHFFFAOYSA-N
120181
C1CNCCN1.O.O.O.O.O.O
C4H22N2O6
MFCD00149389
piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, piperazine hexahydrate, vermyl tn
piperazine;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: PIPERAZINE

RTECSNumber : TM0850000

TSCA : TSCA

SDS Documents