Learn More
Piperazine hexahydrate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131300010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Piperazine hexahydrate | |
142-63-2 | |
87°C | |
98% | |
C4H10N2·6H2O | |
1kg | |
14,7464 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol,practically insoluble in ether | |
C1CNCCN1.O.O.O.O.O.O | |
194.23 | |
194.23 |
98% | |
145.0°C to 156.0°C | |
Authentic | |
Plastic bottle | |
MFCD00149389 | |
23,4 | |
piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn | |
AVRVZRUEXIEGMP-UHFFFAOYSA-N | |
piperazine;hexahydrate | |
120181 | |
98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
RTECSNumber : TM0850000
RUO â Research Use Only