Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperazine dihydrochloride monohydrate, 98%

$61.26 - $151.16

Chemical Identifiers

Chemical Name or Material Piperazine dihydrochloride monohydrate
CAS 6091-62-9
Molecular Formula C4H10N2·2HCl·H2O
Formula Weight 177.08 (159.06 Anhydrous)
MDL Number MFCD00150089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA185200B

 
alfa aesar™
A185200B
1000g Each for $151.16

AAA1852030

 
alfa aesar™
A1852030
250g Each for $61.26
Description & Specifications

Specifications

Formula Weight 177.08 (159.06 Anhydrous)
Chemical Name or Material Piperazine dihydrochloride monohydrate
TSCA TSCA
Assay 98%
Molecular Formula C4H10N2·2HCl·H2O
MDL Number MFCD00150089
CAS 6091-62-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H361-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TL4025000
Odor Odorless
Melting Point >300°C
Quantity 1000g
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H361-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.