Learn More
Piperazine, anhydrous, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1504922
Description
Piperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPiperazine acts as an intermediate for pharmaceuticals, polymers, dyes, corrosion inhibitors, rubber accelerators and surfactants. It is used as a raw material in the manufacture of plastics, polyamide resins, urethane and brake fluid. It is also used for carbondioxide and hydrogen sulfide gas scrubbing mediums. Furthermore, it serves as an epoxy curing agent.
Solubility
Soluble in water, chloroform, glycerol, glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether.
Notes
Air and light sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Specifications
Piperazine | |
110-85-0 | |
1.11 | |
88°C (190°F) | |
MFCD00005953 | |
UN2579 | |
Hygroscopic | |
diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol | |
GLUUGHFHXGJENI-UHFFFAOYSA-N | |
piperazine | |
4837 | |
86.14 |
anhydrous | |
108°C to 113°C | |
146°C to 148°C | |
C4H10N2 | |
100 g | |
102555 | |
14,7464 | |
Soluble in water,chloroform,glycerol,glycols and alcohol. Slightly soluble in benzene and heptane. Insoluble in ether. | |
C1CNCCN1 | |
86.14 | |
CHEBI:28568 | |
99% |
Chemical Identifiers
110-85-0 | |
86.14 | |
GLUUGHFHXGJENI-UHFFFAOYSA-N | |
4837 | |
piperazine |
C4H10N2 | |
MFCD00005953 | |
diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol | |
CHEBI:28568 | |
C1CNCCN1 |
Safety and Handling
GHS H Statement
H334-H361-H314-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
P201-P202-P210-P240-P241-P260-P264b-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
H228-H314-H317-H334-H361fd
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PIPERAZINE
EINECSNumber : 203-808-3
RTECSNumber : TK7800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only