Learn More
Piperazine, 99%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131290025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Piperazine | |
110-85-0 | |
White to Yellow | |
65°C | |
98.5% min. (GC) | |
C4H10N2 | |
2.5 kg | |
04,392 | |
diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol | |
GLUUGHFHXGJENI-UHFFFAOYSA-N | |
piperazine | |
4837 | |
86.14 | |
Extra Pure |
anhydrous, 99% | |
107.0°C to 111.0°C | |
146.0°C to 148.0°C | |
Authentic | |
Plastic drum | |
MFCD00005953 | |
23,4 | |
15,7576 | |
Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol and glycols, soluble in alcohol, practically insoluble in diethyl ether | |
C1CNCCN1 | |
86.14 | |
CHEBI:28568 | |
99% | |
Crystalline Flakes |
Chemical Identifiers
110-85-0 | |
86.14 | |
GLUUGHFHXGJENI-UHFFFAOYSA-N | |
4837 | |
piperazine |
C4H10N2 | |
MFCD00005953 | |
diethylenediamine, piperazin, 1,4-diazacyclohexane, hexahydropyrazine, piperazidine, antiren, 1,4-piperazine, diethyleneimine, eraverm, pipersol | |
CHEBI:28568 | |
C1CNCCN1 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mis
GHS Signal Word: Danger
EINECSNumber : 203-808-3
RTECSNumber : TK7800000
TSCA : TSCA
RUO – Research Use Only