missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Piperazine-2-one, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 398710050

 View more versions of this product

Catalog No. AC398710050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Piperazine-2-one
Authentic
Glass bottle
MFCD01318687
2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine
O=C1C[NH2+]CCN1
101.13
100.12
5625-67-2
94% min. (GC)
C4H9N2O
5g
IWELDVXSEVIIGI-UHFFFAOYSA-O
piperazin-2-one
231360
95%

Chemical Identifiers

5625-67-2
101.13
IWELDVXSEVIIGI-UHFFFAOYSA-O
231360
O=C1C[NH2+]CCN1
C4H9N2O
MFCD01318687
2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine
piperazin-2-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only