Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Piperazine-2-carboxylic acid dihydrochloride, 98%

$87.08 - $281.77

Chemical Identifiers

Chemical Name or Material (+/-)-Piperazine-2-carboxylic acid dihydrochloride
CAS 3022-15-9
Molecular Formula C5H10N2O2·2HCl
Formula Weight 203.07
MDL Number MFCD00044803
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2710106

  SDS
alfa aesar™
H2710106
5g Each for $87.08

AAH2710114

  SDS
alfa aesar™
H2710114
25g Each for $281.77
Description & Specifications

Specifications

Formula Weight 203.07
Chemical Name or Material (+/-)-Piperazine-2-carboxylic acid dihydrochloride
Assay 98%
Molecular Formula C5H10N2O2·2HCl
MDL Number MFCD00044803
CAS 3022-15-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 270° to 273°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.