Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Piperazine-2-carboxylic acid, 98%

$54.39 - $456.55

Chemical Identifiers

Chemical Name or Material (+/-)-Piperazine-2-carboxylic acid
CAS 2762-32-5
Molecular Formula C5H10N2O2
Formula Weight 130.15
MDL Number MFCD00132878
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2734403

  SDS
alfa aesar™
H2734403
1g Each for $54.39

AAH2734406

  SDS
alfa aesar™
H2734406
5g Each for $152.19

AAH2734414

  SDS
alfa aesar™
H2734414
25g Each for $456.55
Description & Specifications

Specifications

Formula Weight 130.15
Health Hazard 1 Warning
Assay 98%
Molecular Formula C5H10N2O2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00132878
Melting Point ≈290°C
CAS 2762-32-5
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (+/-)-Piperazine-2-carboxylic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.