Please login to your online account to display your discounted pricing

Alfa Aesar™ Piperacillin, 95%

$127.62 - $127.62

Chemical Identifiers

Chemical Name or Material Piperacillin
CAS 61477-96-1
Formula Weight 517.55
MDL Number MFCD00865043
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6614303

  SDS
alfa aesar™
J6614303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66143ME

  SDS
alfa aesar™
J66143ME
500mg Each for $127.62
Description & Specifications

Specifications

Formula Weight 517.55
Melting Point 139° to 140°C
Chemical Name or Material Piperacillin
MDL Number MFCD00865043
Assay 95%
CAS 61477-96-1
Health Hazard 1 Danger
Color White
Physical Form Powder
Storage Note 1 2° to 8°C
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Solubility Information Freely soluble in methanol; Only sparingly soluble in aqueous solution at 0.119mg/mL
RTECS Number XH8952200

Applications
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.

Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

2° to 8°C