missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pipemidic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6643709

 View more versions of this product

Catalog No. AAJ6643709



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pipemidic Acid
Powder
251°C to 255°C
MFCD00057291
Soluble in 1N sodium hydroxide or in DMSO. Insoluble in water
CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1
303.32
CHEBI:75250
98%
10 g
Light sensitive
Pipemidic acid
51940-44-4
C14H17N5O3
14,7460
JOHZPMXAZQZXHR-UHFFFAOYSA-N
8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid
4831
303.32
White
>110°C (230°F)
717°C

Chemical Identifiers

51940-44-4
303.32
JOHZPMXAZQZXHR-UHFFFAOYSA-N
CHEBI:75250
CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1
C14H17N5O3
MFCD00057291
4831
8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

EINECSNumber : 257-530-2

RTECSNumber : UV1153800

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only