missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pipamperone, TCI America™

Manufacturer:  TCI America P2315100MG

 View more versions of this product

Catalog No. P2315100MG



Specifications

Specifications

Pipamperone
126°C
1893-33-0
C21H30FN3O2
2811
AXKPFOAXAHJUAG-UHFFFAOYSA-N
1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide
4830
375.49
White-Yellow
100mg
≥98.0% (HPLC)
MFCD00242979
Floropipamide, 1′-[4-(4-Fluorophenyl)-4-oxobutyl]-[1,4′-bipiperidine]-4′-carboxamide
C1CCN(CC1)C2(CCN(CC2)CCCC(=O)C3=CC=C(C=C3)F)C(=O)N
375.488
CHEBI:78549
Crystal-Powder at 20°C

Chemical Identifiers

1893-33-0
375.488
AXKPFOAXAHJUAG-UHFFFAOYSA-N
4830
1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide
C21H30FN3O2
MFCD00242979
Floropipamide, 1′-[4-(4-Fluorophenyl)-4-oxobutyl]-[1,4′-bipiperidine]-4′-carboxamide
CHEBI:78549
C1CCN(CC1)C2(CCN(CC2)CCCC(=O)C3=CC=C(C=C3)F)C(=O)N
Safety and Handling

Safety and Handling

RTECSNumber : DW1050000

TSCA : No

Documents