Please login to your online account to display your discounted pricing

PIP™ TOWA™ PowerGrab™ Latex-Coated Palm Textured Finish Gloves

Ideal for general material handling, assembly, construction and inspection applications

$36.65 - $36.65

Specifications

Cleaning Procedure Recommend washing gloves in slightly warm Water not exceeding 40°C, a mild non-ionic Detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C
Coated Area Palm, finger
Color Blue
Glove Cuff Color Gray
Material Cotton
View More Specs

 Alert

Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Products
Catalog Number Mfr. No. Hem Color Length (English) Width (English) Size Price Quantity    

19-816-905

 
protective industrial
39C1300L
Brown 10.2 in. 4.1 in. Large Dozen of 12 Pair for $36.65
Case of 6 Dozen for $210.84
Description & Specifications

Specifications

Cleaning Procedure Recommend washing gloves in slightly warm Water not exceeding 40°C, a mild non-ionic Detergent should be used, a 5 to 10 min. cycle time should be employed, tumble dry on low maximum temperature 60°C
Coated Area Palm, finger
Color Blue
Glove Cuff Color Gray
Material Cotton
Material Polyester
Exterior Finish Crinkle
For Use With (Application) assembly, construction, inspection, material handling, refuse handling, landscaping and gardening
Glove Cuff Style Knit

Machine washable work glove features blue latex rubber coating on palm and fingertips, and a fully coated thumb. Cuff color denotes size.

  • Crinkle-textured finish for superior grip in wet and dry applications
  • Ergonomic shape for reduced hand fatigue
  • Breathable seamless, gray cotton-polyester blend string knit liner