missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ Temp-Gard™ Extreme Temperature Gloves

With strong grip and flexibility even at extreme temperatures

$134.76 - $240.44

Specifications

Color Black
Material Silicone
For Use With (Application) cryogenic cleaning, cryogenic systems maintenance, conservation of food produce
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Description Length (English) Width (English) Size Price Quantity  
Catalog Number Mfr. No. Description Length (English) Width (English) Size Price Quantity  
19-130-5457 Protective Industrial
2021012L
Wrist length 12.5 in. 5.1 in. Large N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-130-5458 Protective Industrial
2021015S
Mid-arm length 13.5 in. 4.7 in. Small Pair for $174.03
19-130-5459 Protective Industrial
2021015M
Mid-arm length 14.3 in. 4.9 in. Medium Pair for $174.03
19-130-5460 Protective Industrial
2021015L
Mid-arm length 15 in. 5.1 in. Large Pair for $174.03
19-130-5461 Protective Industrial
2021019S
Elbow length 17.25 in. 4.7 in. Small Pair for $198.53
19-130-5462 Protective Industrial
2021019M
Elbow length 18 in. 4.9 in. Medium Pair for $198.53
19-130-5463 Protective Industrial
2021019L
Elbow length 19.25 in. 5.1 in. Large Pair for $198.53
19-130-5464 Protective Industrial
2021027S
Shoulder length 24.75 in. 4.7 in. Small Pair for $240.44
19-130-5465 Protective Industrial
2021027M
Shoulder length 25.5 in. 4.9 in. Medium Pair for $240.44
19-130-5466 Protective Industrial
2021027L
Shoulder length 27 in. 5.1 in. Large Pair for $240.44
19-130-5455 Protective Industrial
2021012S
Wrist length 11.3 in. 4.7 in. Small N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-130-5456 Protective Industrial
2021012M
Wrist length 12 in. 4.9 in. Medium N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Works as cryo gloves and liquid nitrogen gloves
  • Made from liquid-proof silicone fabric
  • Impervious to cryogenic liquids and most other liquids and chemicals
  • Easy to clean, will not absorb chemicals and will not stain
  • Can be used at temperatures ranging from -196° to +260°C (-321° to +500°F)
  • For use in extreme temperature procedures such as cryogenic cleaning, cryogenic systems maintenance and conservation of food produce
  • Available in different sizes of wrist, mid-arm, elbow and shoulder lengths in black color
  • Wrist length gloves come with elastic cuffs and other sizes come with gauntlet cuffs

 

Description & Specifications

Specifications

Black
Silicone
cryogenic cleaning, cryogenic systems maintenance, conservation of food produce