PIP™ Temp-Gard™ Extreme Temperature Apron
Black silicone apron protects torso and upper legs in extreme temperature conditions
$197.96 - $266.44
Specifications
Width (English) 24 in.
Width (Metric) 61cm
View More Specs
Products 4
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity  
19149651 View Documents PIP™
2022042CN
42 in. 106.7cm Each for $220.09
19149650 View Documents PIP™
2022036
36 in. 91.4cm Each for $197.96
19149653 View Documents PIP™
2022054
54 in. 137.2cm Each for $266.44
19149652 View Documents PIP™
2022048CN
48 in. 122cm Each for $243.28
Description
Description
  • Protects from splashes
  • Multi-layered liquid-proof black silicone fabric
  • Impervious to cryogenic liquids and most other liquids and chemicals
  • Does not become slippery, even at extreme temperatures
  • Superior flexibility; will not stiffen
  • Easy to clean; will not absorb chemicals and will not stain
  • Quick-release buckles at neck and waist for added safety
  • Temperature range: -196° to +260°C (-321° to +500°F)
Cryogenic cleaning, cryogenic systems maintenance, conservation of food produce.
Specifications
Specifications
24 in.
61cm