PIP™ Temp-Gard™ Extreme Temperature Apron
Black silicone apron protects torso and upper legs in extreme temperature conditions
Manufacturer:  PIP™ 2022036
 View more versions of this product
Catalog No. 19149650
Description
Description
  • Protects from splashes
  • Multi-layered liquid-proof black silicone fabric
  • Impervious to cryogenic liquids and most other liquids and chemicals
  • Does not become slippery, even at extreme temperatures
  • Superior flexibility; will not stiffen
  • Easy to clean; will not absorb chemicals and will not stain
  • Quick-release buckles at neck and waist for added safety
  • Temperature range: -196° to +260°C (-321° to +500°F)
Cryogenic cleaning, cryogenic systems maintenance, conservation of food produce.
Specifications
Specifications
36 in.
24 in.
36 x 24 in. (91.4 x 61cm)
91.4cm
61cm
Documents