missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PIP™ NOVAX™ Electrical Insulating Gloves

Class 2 electrical rated gloves

$91.20 - $91.20

Specifications

Arc Flash Yes
Certifications ISO9001:2000
Classification 2
Color Black
Length (English) 14 in.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Width (English) Width (Metric) Size Price Quantity  
Catalog Number Mfr. No. Width (English) Width (Metric) Size Price Quantity  
19-168-2124 Protective Industrial
1502148
3.9 in. 10cm 8 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-168-2125 Protective Industrial
1502149
4.1 in. 10.5cm 9 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-168-2126 Protective Industrial
15021410
4.7 in. 12cm 10 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-168-2127 Protective Industrial
15021411
5.3 in. 13.5cm 11 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Natural rubber construction offers the required dielectric properties combined with flexibility, strength and durability.

  • Silicone free
  • Length: 14 in. (35.56cm)
  • Black
  • Unlined
  • Cuff style: rolled
  • Smooth finish

Applications:

Used exclusively for electrical purposes, such as power generation, electrical contractors, hybrid automotive repair/service, hybrid automobile manufacturing, industrial plant maintenance and utilities

Certifications

ISO9001:2000 certified

Compliance

Meet ANSI/ASTM D120 standard and NFPA 70E for use around electrical hazards and arc flash protection

Description & Specifications

Specifications

Yes
2
14 in.
Rubber, natural
Rolled
17000VAC, 25500VDC
ISO9001:2000
Black
35.5cm
exclusively for electrical purposes, such as power generation, electrical contractors, hybrid automotive repair/service, hybrid automobile manufacturing, industrial plant maintenance, utilities
2.29mm